missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[4-(tert-Butyldimethylsilyloxymethyl)-2-nitrophenoxy]acetic Acid 98.0+%, TCI America™
$234.87 - $1266.62
Chemical Identifiers
| CAS | 308815-83-0 |
|---|---|
| Molecular Formula | C15H23NO6Si |
| Molecular Weight (g/mol) | 341.44 |
| MDL Number | MFCD04117911 |
| InChI Key | UXUZQGZIXUXYHB-UHFFFAOYSA-N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B2322100MG
|
TCI America
B2322100MG |
100 mg |
Each for $234.87
|
|
|||||
|
B23221G
|
TCI America
B23221G |
1 g |
Each for $1,266.62
|
|
|||||
Specifications
| 308815-83-0 | |
| C15H23NO6Si | |
| 100 mg | |
| 341.44 | |
| ≥98.0% (HPLC,T) | |
| 2-[4-(tert-Butyldimethylsilyloxymethyl)-2-nitrophenoxy]acetic Acid |
| Yellow | |
| MFCD04117911 | |
| UXUZQGZIXUXYHB-UHFFFAOYSA-N | |
| 341.44 | |
| Crystalline Powder |
Safety and Handling
TSCA : No