missing translation for 'onlineSavingsMsg'
Learn More

2,4-TDI, MilliporeSigma™ Supelco™

Catalog No. 111019247
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019247 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019247 Supplier Merck Emd Millipore Supplier No. 334275MLR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 641-74-7
Density 1.22 g/mL (at 20°C)
Flash Point 132°C - Closed Cup
Physical Form Neat
Linear Formula C9H6N2O2
MDL Number MFCD00002011
Melting Point 19.5°C to 21.5°C
Boiling Point 124°C to 126°C (18 mmHg); 251°C
Synonym Toluene 2,4-diisocyanate; 2,4-Diisocyanatotoluene; 4-Methyl-m-phenylene diisocyanate; Tolylene 2,4-diisocyanate
Type Analytical Standard
Quantity 5 mL
Formula Weight 174.16
Grade Analytical Standard
Show More Show Less