missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Pyridyl)ethanesulfonic Acid 98.0+%, TCI America™

Catalog No. P035325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P035325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P035325G Supplier TCI America Supplier No. P035325G

Chemical Identifiers

CAS 53054-76-5
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00006452
InChI Key RGIIAYDCZSXHGL-UHFFFAOYSA-N
Synonym 4-pyridineethanesulfonic acid, 2-4-pyridyl ethanesulfonic acid, 4-pyridylethylsulfonic acid, 2-pyridin-4-yl ethanesulfonic acid, 4-pyridyl-2-ethyl sulfonic acid, 4-pyridineethanesulfonicacid, 2-4-pyridyl ethanesulphonic acid, 4-2-sulfoethyl pyridine, acmc-209l3n, 4-2-sulfoethyl-pyridine
PubChem CID 96722
IUPAC Name 2-pyridin-4-ylethanesulfonic acid
SMILES C1=CN=CC=C1CCS(=O)(=O)O

Specifications

Melting Point 299°C
Color White-Yellow
Quantity 25 g
UN Number 2585
Formula Weight 187.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Pyridyl)ethanesulfonic Acid
EINECSNumber (5)-5909
TSCA Yes