CAS RN 53054-76-5
IUPAC Name:
2-(pyridin-4-yl)ethane-1-sulfonic acid
Synonyms:
2-(pyridin-4-yl)ethanesulfonic acid
Molecular Weight (g/mol):
187.21
Molecular Formula:
C7H9NO3S
InChi Key:
RGIIAYDCZSXHGL-UHFFFAOYSA-N
SMILES:
OS(=O)(=O)CCC1=CC=NC=C1
1
–
1
of
1
results
1
2-(4-Pyridyl)ethanesulfonic Acid 98.0+%, TCI America™
CAS: 53054-76-5 Molecular Formula: C7H9NO3S Molecular Weight (g/mol): 187.213 MDL Number: MFCD00006452 InChI Key: RGIIAYDCZSXHGL-UHFFFAOYSA-N Synonym: 4-pyridineethanesulfonic acid,2-4-pyridyl ethanesulfonic acid,4-pyridylethylsulfonic acid,2-pyridin-4-yl ethanesulfonic acid,4-pyridyl-2-ethyl sulfonic acid,4-pyridineethanesulfonicacid,2-4-pyridyl ethanesulphonic acid,4-2-sulfoethyl pyridine,acmc-209l3n,4-2-sulfoethyl-pyridine PubChem CID: 96722 IUPAC Name: 2-pyridin-4-ylethanesulfonic acid SMILES: C1=CN=CC=C1CCS(=O)(=O)O