missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Pyridyl)-2-propanol 97.0+%, TCI America™
Supplier: TCI America P147125G
Specifications
| 2-(4-Pyridyl)-2-propanol | |
| 136°C | |
| C8H11NO | |
| 25 g | |
| WEQVCDIJDUVSID-UHFFFAOYSA-N | |
| 2-pyridin-4-ylpropan-2-ol | |
| 4580583 | |
| ≥97.0% (GC,T) |
| 15031-78-4 | |
| 140°C | |
| MFCD00071746 | |
| 2-(4-Pyridyl)isopropyl Alcohol | |
| CC(C)(C1=CC=NC=C1)O | |
| 137.182 | |
| 137.18 | |
| Crystalline Powder |
Chemical Identifiers
| 15031-78-4 | |
| 137.182 | |
| WEQVCDIJDUVSID-UHFFFAOYSA-N | |
| 4580583 | |
| CC(C)(C1=CC=NC=C1)O |
| C8H11NO | |
| MFCD00071746 | |
| 2-(4-Pyridyl)isopropyl Alcohol | |
| 2-pyridin-4-ylpropan-2-ol |
Safety and Handling
TSCA : No