missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Pyridyl)-2-propanol 97.0+%, TCI America™
$102.52 - $303.84
Chemical Identifiers
| CAS | 15031-78-4 |
|---|---|
| Molecular Formula | C8H11NO |
| Molecular Weight (g/mol) | 137.182 |
| MDL Number | MFCD00071746 |
| InChI Key | WEQVCDIJDUVSID-UHFFFAOYSA-N |
| Synonym | 2-(4-Pyridyl)isopropyl Alcohol |
| PubChem CID | 4580583 |
| IUPAC Name | 2-pyridin-4-ylpropan-2-ol |
| SMILES | CC(C)(C1=CC=NC=C1)O |
Chemical Identifiers
| 15031-78-4 | |
| 137.182 | |
| WEQVCDIJDUVSID-UHFFFAOYSA-N | |
| 4580583 | |
| CC(C)(C1=CC=NC=C1)O |
| C8H11NO | |
| MFCD00071746 | |
| 2-(4-Pyridyl)isopropyl Alcohol | |
| 2-pyridin-4-ylpropan-2-ol |
Specifications
| 15031-78-4 | |
| 140°C | |
| MFCD00071746 | |
| 2-(4-Pyridyl)isopropyl Alcohol | |
| CC(C)(C1=CC=NC=C1)O | |
| 137.182 | |
| 137.18 | |
| Crystalline Powder |
| 136°C | |
| C8H11NO | |
| 5 g | |
| WEQVCDIJDUVSID-UHFFFAOYSA-N | |
| 2-pyridin-4-ylpropan-2-ol | |
| 4580583 | |
| ≥97.0% (GC,T) | |
| 2-(4-Pyridyl)-2-propanol |
Safety and Handling
TSCA : No