missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Piperidinyl)benzimidazole 98.0+%, TCI America™

Catalog No. P23821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23821G 1 g
P23825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23821G Supplier TCI America Supplier No. P23821G
Only null left

Chemical Identifiers

CAS 38385-95-4
Molecular Formula C12H15N3
Molecular Weight (g/mol) 201.273
MDL Number MFCD01475843
InChI Key HBOGHPAOOWUTLB-UHFFFAOYSA-N
PubChem CID 715810
IUPAC Name 2-piperidin-4-yl-1H-benzimidazole
SMILES C1CNCCC1C2=NC3=CC=CC=C3N2

Specifications

Melting Point 263°C
Quantity 1 g
Formula Weight 201.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Piperidinyl)benzimidazole