accessibility menu, dialog, popup

UserName

HNNHN

CAS RN 38385-95-4

IUPAC Name: 2-(piperidin-4-yl)-1H-1,3-benzodiazole
Synonyms: 2-(piperidin-4-yl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 201.27
Molecular Formula: C12H15N3
InChi Key: HBOGHPAOOWUTLB-UHFFFAOYSA-N
SMILES: C1CC(CCN1)C1=NC2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results