missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Piperidinedione 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 100°C |
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 113.12 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4-Piperidinedione |
Safety and Handling
| TSCA | No |