missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentanedione, Reagent, 99%, Spectrum™ Chemical

Catalog No. 18600090
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 gal.
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600090 100 mL Amber Glass Bottle
18600091 500 mL Amber Glass Bottle
18600092 1 L Amber Glass Bottle
18600093 5 gal. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600090 Supplier Spectrum Chemical Mfg Cor Supplier No. P1016100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

P1016, 123-54-6, C5H8O2

Spectrum™ Chemical 2,4-Pentanedione, Reagent , also known as acetylacetone, is also a building block for the synthesis of heterocyclic compounds. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O

Specifications

CAS Max % 100%
Boiling Point 140.5°C (284.9°F)
Packaging Amber Glass Bottle
Quantity 100 mL
UN Number UN2310
Percent Purity 99%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2,4-Pentanedione