missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentanedione, 99%

Catalog No. AAA14117AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14117AE 100 mL
AAA14117AP 500 mL
AAA141170F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA14117AE Supplier Thermo Scientific Chemicals Supplier No. A14117AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123-54-6 | C5H8O2 | 100.12 g/mol

2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.

Solubility
It is readily miscible with water.

Notes
Incompatible with isocyanates, aldehydes, cyanides, peroxides, and anhydrides.

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O

Specifications

Melting Point -23°C
Density 0.973
Boiling Point 140°C to 141°C
Flash Point 34°C (93°F)
Assay Percent Range 99%
Refractive Index 1.452
Quantity 100 mL
UN Number UN2310
Beilstein 741937
Merck Index 14,81
Solubility Information It is readily miscible with water.
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2,4-Pentanedione
Show More Show Less
Hazard Category H226-H302-H311+H331
Hazard Statement GHS H Statement
H311-H331-H226-H302
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANE-2,4-DIONE
EINECSNumber 204-634-0
RTECSNumber SA1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only