missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2,4-Pentanedionato)lithium 98.0+%, TCI America™
Supplier: TCI America L008025G
Description
Greener Choice Claims
- Environmental benefits include:
- Source Reduction
Specifications
(2,4-Pentanedionato)lithium | |
250°C | |
C5H7LiO2 | |
25 g | |
JTEOOCAGEXVCBQ-LNKPDPKZSA-M | |
lithium(1+) (2Z)-4-oxopent-2-en-2-olate | |
106.05 | |
Crystalline Powder |
18115-70-3 | |
White-Yellow | |
MFCD00013496 | |
Acetylacetone Lithium Salt, Lithium Acetylacetonate | |
[Li+].C\C([O-])=C\C(C)=O | |
106.05 | |
≥98.0% (T) |
Safety and Handling
TSCA : No