missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentanediol 98.0+%, TCI America™

Supplier:  TCI America P0670500G

 View more versions of this product

Catalog No. P0670500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,4-Pentanediol
Yellow
C5H12O2
500 g
GTCCGKPBSJZVRZ-UHFFFAOYSA-N
pentane-2,4-diol
12262
≥98.0% (GC)
625-69-4
198°C
MFCD00004549
2,4-pentanediol, 2,4-amylene glycol, isoamylene alcohol, pentanediol-2,4, 2,4-amyleneglycol, dl-2,4-pentanediol, 2,4-pentanediol dl+meso, wln: qy&1yq, 2,4-pentandiol, 2,4-dihydroxypentane
CC(CC(C)O)O
104.149
104.15
Liquid

Chemical Identifiers

625-69-4
104.149
GTCCGKPBSJZVRZ-UHFFFAOYSA-N
12262
CC(CC(C)O)O
C5H12O2
MFCD00004549
2,4-pentanediol, 2,4-amylene glycol, isoamylene alcohol, pentanediol-2,4, 2,4-amyleneglycol, dl-2,4-pentanediol, 2,4-pentanediol dl+meso, wln: qy&1yq, 2,4-pentandiol, 2,4-dihydroxypentane
pentane-2,4-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0240

RTECSNumber : SA0490000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions