missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentanediol 98.0+%, TCI America™

Catalog No. p-7134722
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P067025G 25 g
P0670500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P067025G Supplier TCI America Supplier No. P067025G
Only null left

Chemical Identifiers

CAS 625-69-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00004549
InChI Key GTCCGKPBSJZVRZ-UHFFFAOYSA-N
Synonym 2,4-pentanediol, 2,4-amylene glycol, isoamylene alcohol, pentanediol-2,4, 2,4-amyleneglycol, dl-2,4-pentanediol, 2,4-pentanediol dl+meso, wln: qy&1yq, 2,4-pentandiol, 2,4-dihydroxypentane
PubChem CID 12262
IUPAC Name pentane-2,4-diol
SMILES CC(CC(C)O)O

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 g
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Pentanediol
EINECSNumber (2)-0240
RTECSNumber SA0490000
TSCA No