Learn More
2-(4-Morpholinyl)ethyl isothiocyanate, 97%, Thermo Scientific Chemicals
$143.34 - $143.34
Chemical Identifiers
| CAS | 63224-35-1 |
|---|---|
| Molecular Formula | C7H12N2OS |
| Molecular Weight (g/mol) | 172.25 |
| MDL Number | MFCD00041221 |
| InChI Key | SDNSCXCXVQHBGH-UHFFFAOYSA-N |
| Synonym | 2-morpholinoethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine, 2-4-morpholino ethyl isothiocyanate, 2-morpholinoethylisothiocyanate, 4-2-isothiocyanato-ethyl-morpholine, 2-morpholin-4-ylethanisothiocyanate, 2-4-morpholinyl ethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine # |
| PubChem CID | 143957 |
| IUPAC Name | 4-(2-isothiocyanatoethyl)morpholine |
| SMILES | S=C=NCCN1CCOCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1204604
|
Thermo Scientific Chemicals
L1204604 |
2 g |
Each for $143.34
|
|
|||||
Description
2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.
Solubility
Hydrolyzes in water.
Notes
Store at 4°C. Store under dry inert gas. Protect from humidity and water. Incompatible with moisture, amines, alcohols and oxidizing agents. Protect from heat. Moisture Sensitive.
Chemical Identifiers
| 63224-35-1 | |
| 172.25 | |
| SDNSCXCXVQHBGH-UHFFFAOYSA-N | |
| 143957 | |
| S=C=NCCN1CCOCC1 |
| C7H12N2OS | |
| MFCD00041221 | |
| 2-morpholinoethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine, 2-4-morpholino ethyl isothiocyanate, 2-morpholinoethylisothiocyanate, 4-2-isothiocyanato-ethyl-morpholine, 2-morpholin-4-ylethanisothiocyanate, 2-4-morpholinyl ethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine # | |
| 4-(2-isothiocyanatoethyl)morpholine |
Specifications
| 63224-35-1 | |
| 162°C to 163°C (10 mmHg) | |
| C7H12N2OS | |
| MFCD00041221 | |
| UN2922 | |
| Moisture sensitive | |
| Hydrolyzes in water. | |
| S=C=NCCN1CCOCC1 | |
| 172.25 | |
| 172.25 | |
| 2-(4-Morpholinyl)ethyl isothiocyanate |
| 1.11 | |
| >110°C (230°F) | |
| 1.539 | |
| 2 g | |
| 1210530 | |
| 2-morpholinoethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine, 2-4-morpholino ethyl isothiocyanate, 2-morpholinoethylisothiocyanate, 4-2-isothiocyanato-ethyl-morpholine, 2-morpholin-4-ylethanisothiocyanate, 2-4-morpholinyl ethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine # | |
| SDNSCXCXVQHBGH-UHFFFAOYSA-N | |
| 4-(2-isothiocyanatoethyl)morpholine | |
| 143957 | |
| 97% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only