missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylpiperazin-1-yl)benzoic acid, ≥97%, Thermo Scientific™

$148.38 - $148.38

Chemical Identifiers

CAS 159589-70-5
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD01788118
InChI Key WKGFDTBUUBBWJZ-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-yl benzoic acid, 2-4-methyl-piperazin-1-yl-benzoic acid, 2-4-methyl-1-piperazinyl benzoic acid, 1-2-carboxyphenyl-4-methylpiperazine, 2-4-methylpiperazinyl benzoic acid, 4-methyl-piperazin-1-yl-benzoic acid, 2-4-methylpiperazin-1-yl benzoicacid, benzoic acid,2-4-methyl-1-piperazinyl, benzoic acid, 2-4-methyl-1-piperazinyl
PubChem CID 1120459
IUPAC Name 2-(4-methylpiperazin-1-yl)benzoic acid
SMILES CN1CCN(CC1)C1=CC=CC=C1C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC36001CB
SDS View Documents
Thermo Scientific Chemicals
CC36001CB
250 mg Amber Glass Bottle
Each for $148.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

159589-70-5
220.27
WKGFDTBUUBBWJZ-UHFFFAOYSA-N
1120459
CN1CCN(CC1)C1=CC=CC=C1C(O)=O
C12H16N2O2
MFCD01788118
2-4-methylpiperazin-1-yl benzoic acid, 2-4-methyl-piperazin-1-yl-benzoic acid, 2-4-methyl-1-piperazinyl benzoic acid, 1-2-carboxyphenyl-4-methylpiperazine, 2-4-methylpiperazinyl benzoic acid, 4-methyl-piperazin-1-yl-benzoic acid, 2-4-methylpiperazin-1-yl benzoicacid, benzoic acid,2-4-methyl-1-piperazinyl, benzoic acid, 2-4-methyl-1-piperazinyl
2-(4-methylpiperazin-1-yl)benzoic acid

Specifications

159589-70-5
100.0
Amber Glass Bottle
MFCD01788118
2-4-methylpiperazin-1-yl benzoic acid, 2-4-methyl-piperazin-1-yl-benzoic acid, 2-4-methyl-1-piperazinyl benzoic acid, 1-2-carboxyphenyl-4-methylpiperazine, 2-4-methylpiperazinyl benzoic acid, 4-methyl-piperazin-1-yl-benzoic acid, 2-4-methylpiperazin-1-yl benzoicacid, benzoic acid,2-4-methyl-1-piperazinyl, benzoic acid, 2-4-methyl-1-piperazinyl
CN1CCN(CC1)C1=CC=CC=C1C(O)=O
220.27
220.27
2-(4-Methylpiperazin-1-yl)benzoic acid
96.5
97%
C12H16N2O2
250 mg
WKGFDTBUUBBWJZ-UHFFFAOYSA-N
2-(4-methylpiperazin-1-yl)benzoic acid
1120459
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'