missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-5-thiazolyl)ethyl Butyrate 98.0+%, TCI America™

Catalog No. p-7133249
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24895G 5 g
M248925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24895G Supplier TCI America Supplier No. M24895G

Chemical Identifiers

CAS 94159-31-6
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.30
MDL Number MFCD09032913
InChI Key GDRZNYCKSKHESZ-UHFFFAOYSA-N
Synonym Butyric Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Butyryloxyethyl)-4-methylthiazole
PubChem CID 3023838
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl butanoate
SMILES CCCC(=O)OCCC1=C(C)N=CS1

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 213.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Methyl-5-thiazolyl)ethyl Butyrate
EINECSNumber (5)-3412
TSCA No