missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Methyl-5-thiazolyl)ethyl Acetate 98.0+%, TCI America™
Supplier: TCI America M18485G
Specifications
| 2-(4-Methyl-5-thiazolyl)ethyl Acetate | |
| Yellow | |
| C8H11NO2S | |
| 5 g | |
| CRTCWNPLKVVXIX-UHFFFAOYSA-N | |
| 2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate | |
| 61192 | |
| ≥98.0% (GC) |
| 656-53-1 | |
| 118°C | |
| MFCD00005338 | |
| 2-4-methylthiazol-5-yl ethyl acetate, 4-methyl-5-thiazolylethyl acetate, 2-4-methyl-5-thiazolyl ethyl acetate, sulfurol acetate, 4-methyl-5-2-acetoxyethyl thiazole, 4-methyl-5-thiazoleethanol acetate, 5-2-acetoxyethyl-4-methylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethyl acetate, 5-thiazoleethanol, 4-methyl-, acetate ester, 4-methyl-5-beta-acetoxyethyl thiazole | |
| CC(=O)OCCC1=C(C)N=CS1 | |
| 185.24 | |
| 185.24 | |
| Liquid |
Chemical Identifiers
| 656-53-1 | |
| 185.24 | |
| CRTCWNPLKVVXIX-UHFFFAOYSA-N | |
| 61192 | |
| CC(=O)OCCC1=C(C)N=CS1 |
| C8H11NO2S | |
| MFCD00005338 | |
| 2-4-methylthiazol-5-yl ethyl acetate, 4-methyl-5-thiazolylethyl acetate, 2-4-methyl-5-thiazolyl ethyl acetate, sulfurol acetate, 4-methyl-5-2-acetoxyethyl thiazole, 4-methyl-5-thiazoleethanol acetate, 5-2-acetoxyethyl-4-methylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethyl acetate, 5-thiazoleethanol, 4-methyl-, acetate ester, 4-methyl-5-beta-acetoxyethyl thiazole | |
| 2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate |
Safety and Handling
EINECSNumber : (5)-6975
TSCA : Yes