missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-5-thiazolyl)ethyl Acetate 98.0+%, TCI America™

Catalog No. p-7132878
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18485G 5 g
M184825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18485G Supplier TCI America Supplier No. M18485G
Only null left

Chemical Identifiers

CAS 656-53-1
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.24
MDL Number MFCD00005338
InChI Key CRTCWNPLKVVXIX-UHFFFAOYSA-N
Synonym 2-4-methylthiazol-5-yl ethyl acetate, 4-methyl-5-thiazolylethyl acetate, 2-4-methyl-5-thiazolyl ethyl acetate, sulfurol acetate, 4-methyl-5-2-acetoxyethyl thiazole, 4-methyl-5-thiazoleethanol acetate, 5-2-acetoxyethyl-4-methylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethyl acetate, 5-thiazoleethanol, 4-methyl-, acetate ester, 4-methyl-5-beta-acetoxyethyl thiazole
PubChem CID 61192
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate
SMILES CC(=O)OCCC1=C(C)N=CS1

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
Formula Weight 185.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Methyl-5-thiazolyl)ethyl Acetate
EINECSNumber (5)-6975
TSCA Yes