missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)ethanol, 98%

Catalog No. AAL0615809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0615809 10 g
AAL0615818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0615809 Supplier Thermo Scientific Chemicals Supplier No. L0615809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 702-23-8 | C9H12O2 | 152.193 g/mol

2-(4-Methoxyphenyl)ethanol is used as an internal standard in the fluorous biphasic catalysis reaction. It is used in the preparation of 4-(2-iodoethyl)phenol, by refluxing it with 47% hydriodic acid. It may be used in the preparation of (2R*,4R*)-1-n-butyl-2-methyl-4-(2-oxopyrrolidin-1-yl)-6-methoxy-1,2,3,4-tetrahydroquinoline and (2R*,4S*)-1-n-butyl-2-methyl-4-(2-oxopyrrolidin-1-yl)-6-methoxy-1,2,3,4- tetrahydroquinoline. It is used for foods, perfumes, and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Methoxyphenyl)ethanol is used as an internal standard in the fluorous biphasic catalysis reaction. It is used in the preparation of 4-(2-iodoethyl)phenol, by refluxing it with 47% hydriodic acid. It may be used in the preparation of (2R*,4R*)-1-n-butyl-2-methyl-4-(2-oxopyrrolidin-1-yl)-6-methoxy-1,2,3,4-tetrahydroquinoline and (2R*,4S*)-1-n-butyl-2-methyl-4-(2-oxopyrrolidin-1-yl)-6-methoxy-1,2,3,4- tetrahydroquinoline. It is used for foods, perfumes, and cosmetics.

Solubility
Soluble in DMSO, Methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at 4°C.

Chemical Identifiers

CAS 702-23-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002900
InChI Key AUWDOZOUJWEPBA-UHFFFAOYSA-N
Synonym 2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol
PubChem CID 69705
IUPAC Name 2-(4-methoxyphenyl)ethanol
SMILES COC1=CC=C(C=C1)CCO

Specifications

Melting Point 26°C to 30°C
Boiling Point 334°C to 336°C
Flash Point >110°C (230°F)
Refractive Index 1.537
Quantity 10 g
Beilstein 2043563
Solubility Information Soluble in DMSO,Methanol.
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-(4-Methoxyphenyl)ethanol
EINECSNumber 211-866-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only