missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxycyclohexyl)ethylamine (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. M23815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23815G 5 g
M23811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23815G Supplier TCI America Supplier No. M23815G

Chemical Identifiers

CAS 1052223-70-7
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.257
MDL Number MFCD07772981
InChI Key CDTAOSZEZXQTRJ-UHFFFAOYSA-N
Synonym 1-(2-Aminoethyl)-4-methoxycyclohexane
PubChem CID 53434199
IUPAC Name 2-(4-methoxycyclohexyl)ethanamine
SMILES COC1CCC(CC1)CCN

Specifications

Color Colorless
Quantity 5 g
UN Number 2735
Formula Weight 157.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(4-Methoxycyclohexyl)ethylamine (cis- and trans- mixture)