accessibility menu, dialog, popup

UserName

H3CONH2

CAS RN 1052223-70-7

IUPAC Name: 2-(4-methoxycyclohexyl)ethan-1-amine
Synonyms: 2-(4-methoxycyclohexyl)ethanamine
Molecular Weight (g/mol): 157.26
Molecular Formula: C9H19NO
InChi Key: CDTAOSZEZXQTRJ-UHFFFAOYSA-N
SMILES: COC1CCC(CCN)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results