missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium Chloride 98.0+%, TCI America™

Catalog No. I00671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00671G 1 g
I00675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00671G Supplier TCI America Supplier No. I00671G

Chemical Identifiers

CAS 146-68-9
Molecular Formula C19H13ClIN5O2
Molecular Weight (g/mol) 505.70
MDL Number MFCD00011961,MFCD00149999
InChI Key JORABGDXCIBAFL-UHFFFAOYSA-M
Synonym iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride
PubChem CID 64957
ChEBI CHEBI:75421
IUPAC Name 3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 505.70
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium Chloride
TSCA Yes
Recommended Storage Refrigerator