missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)propionic acid, 98%

Catalog No. aab2493818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2493818 50 g
AAB2493809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2493818 Supplier Thermo Scientific Chemicals Supplier No. B2493818
Only null left

CAS: 938-96-5 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 938-96-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00128847
InChI Key ZHMMPVANGNPCBW-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenyl propanoic acid, 2-4-hydroxyphenyl propionic acid, 4-hydroxyhydratropate, 4-hydroxyhydratropic acid, 4-hydroxyphenyl-2-propionic acid, 2-4'-hydroxyphenyl-propionic acid, 2-4-hydroxyphenyl propionate, acmc-209ro0, 4-hydroxyphenyl propanoic acid, methyl 4-hydroxyphenylacetic acid
PubChem CID 102526
ChEBI CHEBI:1868
IUPAC Name 2-(4-hydroxyphenyl)propanoic acid
SMILES CC(C1=CC=C(C=C1)O)C(=O)O

Specifications

Melting Point 130°C to 132°C
Quantity 50 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-(4-Hydroxyphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-352-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only