missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)ethanol, 98%

Catalog No. AAA1489103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1489103 1 g
AAA1489106 5 g
AAA1489114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1489103 Supplier Thermo Scientific Chemicals Supplier No. A1489103

CAS: 501-94-0 | C8H10O2 | 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 501-94-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002902
InChI Key YCCILVSKPBXVIP-UHFFFAOYSA-N
Synonym tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
PubChem CID 10393
ChEBI CHEBI:1879
IUPAC Name 4-(2-hydroxyethyl)phenol
SMILES C1=CC(=CC=C1CCO)O

Specifications

Melting Point 89°C to 92°C
Boiling Point 139°C (1 mmHg)
Quantity 1 g
Beilstein 1859884
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 2-(4-Hydroxyphenyl)ethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-930-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only