missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiyn-1,6-diol, 97%

Catalog No. p-7058332
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0957603 1 g
AAL0957606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0957603 Supplier Thermo Scientific Chemicals Supplier No. L0957603
Only null left

CAS: 3031-68-3 | C6H6O2 | 110.112 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3031-68-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00020619
InChI Key JXMQYKBAZRDVTC-UHFFFAOYSA-N
Synonym 2,4-hexadiyne-1,6-diol, diacetylene glycol, 2,4-hexadiynediol, 2,4-hexadiyn-1,6-diol, 2,6-diol, acmc-1ahii, hoch2c=c-c=cch2oh, 1,6-dihydroxy-2,4-hexadiyne, hexa-2, 4-diyne-1, 6-diol, jxmqykbazrdvtc-uhfffaoysa
PubChem CID 94973
IUPAC Name hexa-2,4-diyne-1,6-diol
SMILES C(C#CC#CCO)O

Specifications

Melting Point 110°C to 114°C
Linear Formula HOCH2C≡C-C≡CCH2OH
Quantity 1 g
UN Number UN1325
Beilstein 773791
Formula Weight 110.11
Percent Purity 97%
Chemical Name or Material 2,4-Hexadiyn-1,6-diol
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 221-210-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only