missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiene (mixture of isomers) 95.0+%, TCI America™

Catalog No. p-7130493
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03365ML 5 mL
H033625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03365ML Supplier TCI America Supplier No. H03365ML
Only null left

Chemical Identifiers

CAS 592-46-1
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00009297,MFCD00009297
InChI Key APPOKADJQUIAHP-CIIODKQPSA-N
Synonym unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
PubChem CID 643786
IUPAC Name (2Z,4E)-hexa-2,4-diene
SMILES C\C=C\C=C/C

Specifications

Color Yellow
Boiling Point 83°C
Quantity 5 mL
UN Number 2458
Formula Weight 82.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Hexadiene (mixture of isomers)