accessibility menu, dialog, popup

UserName

CH3H3C(E)(Z)

CAS RN 592-46-1

IUPAC Name: (2Z,4E)-hexa-2,4-diene
Synonyms: 2,4-hexadiene (cis,trans)
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: APPOKADJQUIAHP-CIIODKQPSA-N
SMILES: C\C=C\C=C/C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)