missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadienal 95.0+%, TCI America™

Supplier:  TCI America H058125ML

Catalog No. H058125ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,4-Hexadienal
Yellow
MFCD00007004
2922
BATOPAZDIZEVQF-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienal
637564
96.13
Liquid
142-83-6
C6H8O
25 mL
2,4-hexadienal, hexa-2,4-dienal, sorbaldehyde, sorbic aldehyde, trans,trans-2,4-hexadienal, 2e,4e-hexa-2,4-dienal, 2,4-hexadienal, 2e,4e, 2,4-hexadienal, e,e, 2-propyleneacrolein, 3-propyleneacrolein
CC=CC=CC=O
96.129
CHEBI:82334
≥95.0% (GC)

Chemical Identifiers

142-83-6
96.129
BATOPAZDIZEVQF-MQQKCMAXSA-N
637564
(2E,4E)-hexa-2,4-dienal
C6H8O
MFCD00007004
2,4-hexadienal, hexa-2,4-dienal, sorbaldehyde, sorbic aldehyde, trans,trans-2,4-hexadienal, 2e,4e-hexa-2,4-dienal, 2,4-hexadienal, 2e,4e, 2,4-hexadienal, e,e, 2-propyleneacrolein, 3-propyleneacrolein
CHEBI:82334
CC=CC=CC=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : WG1925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions