accessibility menu, dialog, popup

UserName

CAS RN 142-83-6

IUPAC Name: (2E,4E)-hexa-2,4-dienal
Synonyms: 2,4-hexadienal sorbaldehyde sorbic aldehyde
Molecular Weight (g/mol): 96.13
Molecular Formula: C6H8O
InChi Key: BATOPAZDIZEVQF-MQQKCMAXSA-N
SMILES: C\C=C\C=C\C=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)