missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadienal, predominantly trans,trans, 95%

Catalog No. AAA1320022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1320022 100 g
AAA1320006 5 g
AAA1320014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1320022 Supplier Thermo Scientific Chemicals Supplier No. A1320022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 142-83-6 | C6H8O | 96.129 g/mol

2,4-Hexadienal, is used in the synthesis of chiral cycloadducts. 2,4-Hexadienal is used as a flavouring agent, as a chemical intermediate in various organic synthetic reactions and as a raw material in the manufacture of sorbic acid. It is also used as in the manufacture of polymethine dyes, as a pharmaceutical intermediate in the manufacture of mitomycins and antihypercholesteraemics, as an inhibitor of corrosion for steel used in oil field operations, as a monomer in reactions with silane comonomers for the manufacture of polyalkenyloxysilane polymer and as a fumigant against larvae of the Caribbean fruit fly.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Hexadienal, is used in the synthesis of chiral cycloadducts. 2,4-Hexadienal is used as a flavouring agent, as a chemical intermediate in various organic synthetic reactions and as a raw material in the manufacture of sorbic acid. It is also used as in the manufacture of polymethine dyes, as a pharmaceutical intermediate in the manufacture of mitomycins and antihypercholesteraemics, as an inhibitor of corrosion for steel used in oil field operations, as a monomer in reactions with silane comonomers for the manufacture of polyalkenyloxysilane polymer and as a fumigant against larvae of the Caribbean fruit fly.

Solubility
It is Insoluble in water, but soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Store under inert gas.Take measures to prevent the build up of electrostatic charge. Incompatible with strong reducing agents, strong bases, strong oxidizing agents. It is Stable, Combustible.

Chemical Identifiers

CAS 142-83-6
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00007004
InChI Key BATOPAZDIZEVQF-MQQKCMAXSA-N
Synonym 2,4-hexadienal, hexa-2,4-dienal, sorbaldehyde, sorbic aldehyde, trans,trans-2,4-hexadienal, 2e,4e-hexa-2,4-dienal, 2,4-hexadienal, 2e,4e, 2,4-hexadienal, e,e, 2-propyleneacrolein, 3-propyleneacrolein
PubChem CID 637564
ChEBI CHEBI:82334
IUPAC Name (2E,4E)-hexa-2,4-dienal
SMILES CC=CC=CC=O

Specifications

Density 0.871
Boiling Point 170°C to 173°C
Flash Point 67°C (152°F)
Odor Floral
Refractive Index 1.5386
Quantity 100 g
UN Number UN2922
Beilstein 1698401
Sensitivity Air sensitive
Solubility Information It is Insoluble in water,but soluble in ethanol.
Formula Weight 96.13
Percent Purity 95%
Chemical Name or Material 2,4-Hexadienal, predominantly trans,trans
Show More Show Less
Hazard Category H227-H302-H311-H315-H319-H351
Hazard Statement GHS H Statement
H301-H311-H314-H318-H227-H317
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 205-564-3
RTECSNumber WG1925000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only