missing translation for 'onlineSavingsMsg'
Learn More

2,4-Heptadien-6-one (stabilized with HQ) 95.0+%, TCI America™

Catalog No. h00225g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H00225G Supplier TCI America Supplier No. H00225G

Chemical Identifiers

CAS 3916-64-1
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00059238
InChI Key SWGLACWOVFCDQS-VNKDHWASSA-N
Synonym 5-Acetyl-2,4-pentadiene
PubChem CID 643537
IUPAC Name (3E,5E)-hepta-3,5-dien-2-one
SMILES CC=CC=CC(=O)C

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
Formula Weight 110.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Heptadien-6-one (stabilized with HQ)
RTECSNumber MI5495000
TSCA No