accessibility menu, dialog, popup

UserName

CH3CH3O(E)(E)

CAS RN 3916-64-1

IUPAC Name: (3E,5E)-hepta-3,5-dien-2-one
Synonyms: (3E,5E)-hepta-3,5-dien-2-one
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: SWGLACWOVFCDQS-VNKDHWASSA-N
SMILES: C\C=C\C=C\C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results