missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)indole, 99%

Catalog No. AAA1325503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1325503 1 g
AAA1325506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1325503 Supplier Thermo Scientific Chemicals Supplier No. A1325503

CAS: 782-17-2 | C14H10FN | 211.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 782-17-2
Molecular Formula C14H10FN
Molecular Weight (g/mol) 211.239
MDL Number MFCD00068178
InChI Key VLHGDCJIDNVRFM-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl indole, 2-4-fluorophenyl-1h-indole, pubchem7339, acmc-209pdr, maybridge1_007619, 2-4-fluoro-phenyl-1h-indole
PubChem CID 136622
IUPAC Name 2-(4-fluorophenyl)-1H-indole
SMILES C1=CC=C2C(=C1)C=C(N2)C3=CC=C(C=C3)F

Specifications

Melting Point 186°C to 191°C
Odor Odorless
Quantity 1 g
Beilstein 744766
Formula Weight 211.24
Percent Purity 99%
Chemical Name or Material 2-(4-Fluorophenyl)indole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only