accessibility menu, dialog, popup

UserName

FHN

CAS RN 782-17-2

IUPAC Name: 2-(4-fluorophenyl)-1H-indole
Synonyms: 2-(4-fluorophenyl)-1H-indole
Molecular Weight (g/mol): 211.24
Molecular Formula: C14H10FN
InChi Key: VLHGDCJIDNVRFM-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)