missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Fluorophenyl)ethyl Bromide 95.0+%, TCI America™
$65.24 - $216.24
Chemical Identifiers
| CAS | 332-42-3 |
|---|---|
| Molecular Formula | C8H8BrF |
| Molecular Weight (g/mol) | 203.05 |
| MDL Number | MFCD03844794 |
| InChI Key | FLRUNCJXOVYWDH-UHFFFAOYSA-N |
| Synonym | 1-2-bromoethyl-4-fluorobenzene, 1-fluoro-4-2-bromoethyl benzene, 4-fluorophenethyl bromide, 2-4-fluorophenyl ethyl bromide, 1-bromo-2-4-fluorophenyl ethane, 2-4-fluorophenyl ethylbromide, 4-fluorophenylethyl bromide, 1-2-bromo-ethyl-4-fluoro-benzene, benzene, 1-2-bromoethyl-4-fluoro, 1-2-bromoethyl-4-fluoro-benzene |
| PubChem CID | 573153 |
| IUPAC Name | 1-(2-bromoethyl)-4-fluorobenzene |
| SMILES | FC1=CC=C(CCBr)C=C1 |
Chemical Identifiers
| 332-42-3 | |
| 203.05 | |
| FLRUNCJXOVYWDH-UHFFFAOYSA-N | |
| 573153 | |
| FC1=CC=C(CCBr)C=C1 |
| C8H8BrF | |
| MFCD03844794 | |
| 1-2-bromoethyl-4-fluorobenzene, 1-fluoro-4-2-bromoethyl benzene, 4-fluorophenethyl bromide, 2-4-fluorophenyl ethyl bromide, 1-bromo-2-4-fluorophenyl ethane, 2-4-fluorophenyl ethylbromide, 4-fluorophenylethyl bromide, 1-2-bromo-ethyl-4-fluoro-benzene, benzene, 1-2-bromoethyl-4-fluoro, 1-2-bromoethyl-4-fluoro-benzene | |
| 1-(2-bromoethyl)-4-fluorobenzene |
Specifications
| 332-42-3 | |
| 102°C | |
| MFCD03844794 | |
| 1-2-bromoethyl-4-fluorobenzene, 1-fluoro-4-2-bromoethyl benzene, 4-fluorophenethyl bromide, 2-4-fluorophenyl ethyl bromide, 1-bromo-2-4-fluorophenyl ethane, 2-4-fluorophenyl ethylbromide, 4-fluorophenylethyl bromide, 1-2-bromo-ethyl-4-fluoro-benzene, benzene, 1-2-bromoethyl-4-fluoro, 1-2-bromoethyl-4-fluoro-benzene | |
| FC1=CC=C(CCBr)C=C1 | |
| 203.05 | |
| 203.05 | |
| Liquid |
| Yellow | |
| C8H8BrF | |
| 1 g | |
| FLRUNCJXOVYWDH-UHFFFAOYSA-N | |
| 1-(2-bromoethyl)-4-fluorobenzene | |
| 573153 | |
| ≥95.0% (GC) | |
| 2-(4-Fluorophenyl)ethyl Bromide |
Safety and Handling
TSCA : No