missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)ethyl Bromide 95.0+%, TCI America™

Catalog No. F04311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04311G 1 g
F04315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04311G Supplier TCI America Supplier No. F04311G
Only null left

Chemical Identifiers

CAS 332-42-3
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
MDL Number MFCD03844794
InChI Key FLRUNCJXOVYWDH-UHFFFAOYSA-N
Synonym 1-2-bromoethyl-4-fluorobenzene, 1-fluoro-4-2-bromoethyl benzene, 4-fluorophenethyl bromide, 2-4-fluorophenyl ethyl bromide, 1-bromo-2-4-fluorophenyl ethane, 2-4-fluorophenyl ethylbromide, 4-fluorophenylethyl bromide, 1-2-bromo-ethyl-4-fluoro-benzene, benzene, 1-2-bromoethyl-4-fluoro, 1-2-bromoethyl-4-fluoro-benzene
PubChem CID 573153
IUPAC Name 1-(2-bromoethyl)-4-fluorobenzene
SMILES FC1=CC=C(CCBr)C=C1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 1 g
Formula Weight 203.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Fluorophenyl)ethyl Bromide