Learn More
2-(4-Fluorophenyl)ethanol, 97%
CAS: 7589-27-7 | C8H9FO | 140.16 g/mol
Supplier: Thermo Scientific Chemicals L0561814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-(4-Fluorophenyl)ethanol | |
| 1.121 | |
| 104°C (219°F) | |
| 1.5065 | |
| 25 g | |
| 2-4-fluorophenyl ethanol, 4-fluorophenethyl alcohol, 4-fluorophenylethanol, benzeneethanol, 4-fluoro, 2-4-fluorophenyl ethan-1-ol, 4-fluorophenethylic alcohol, phenethyl alcohol, p-fluoro, 4-fluorobenzeneethanol, 4fa | |
| OCCC1=CC=C(F)C=C1 | |
| 140.16 | |
| 140.16 |
| 7589-27-7 | |
| 118°C (20 mmHg) | |
| C8H9FO | |
| MFCD00002898 | |
| 2354310 | |
| MWUVGXCUHWKQJE-UHFFFAOYSA-N | |
| 2-(4-fluorophenyl)ethanol | |
| 82068 | |
| 97% |
Chemical Identifiers
| 7589-27-7 | |
| 140.16 | |
| MWUVGXCUHWKQJE-UHFFFAOYSA-N | |
| 82068 | |
| OCCC1=CC=C(F)C=C1 |
| C8H9FO | |
| MFCD00002898 | |
| 2-4-fluorophenyl ethanol, 4-fluorophenethyl alcohol, 4-fluorophenylethanol, benzeneethanol, 4-fluoro, 2-4-fluorophenyl ethan-1-ol, 4-fluorophenethylic alcohol, phenethyl alcohol, p-fluoro, 4-fluorobenzeneethanol, 4fa | |
| 2-(4-fluorophenyl)ethanol |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 231-499-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only