missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenoxy)benzaldehyde, 97%

Catalog No. p-7032696
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34259MD 250 mg
AAH3425903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34259MD Supplier Thermo Scientific Chemicals Supplier No. H34259MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 320423-61-8 | C13H9FO2 | 216.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 320423-61-8
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.211
MDL Number MFCD01568770
InChI Key RZJFZXRBXCJPTA-UHFFFAOYSA-N
Synonym 2-4-fluorophenoxy benzaldehyde, 2-4-fluorophenoxy benzenecarbaldehyde, 4-fluorophenoxybenzaldehyde, fluorophenoxybenzenecarbaldehyde, benzaldehyde,2-4-fluorophenoxy
PubChem CID 2782988
IUPAC Name 2-(4-fluorophenoxy)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)OC2=CC=C(C=C2)F

Specifications

Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 216.21
Percent Purity 97%
Chemical Name or Material 2-(4-Fluorophenoxy)benzaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only