missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Ethoxyphenyl)ethanol, 98%

Catalog No. AAL0559704
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0559704 2 g
AAL0559709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0559704 Supplier Thermo Scientific Chemicals Supplier No. L0559704

CAS: 22545-15-9 | C10H14O2 | 166.22 g/mol

2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 22545-15-9
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00016570
InChI Key CNMVSNTVPZWQMI-UHFFFAOYSA-N
Synonym 2-4-ethoxyphenyl ethanol, p-ethoxyphenethyl alcohol, 4-2-hydroxyethyl phenetol, 4-ethoxybenzeneethanol, 4-ethoxy-benzeneethanol, 4-ethoxyphenethyl alcohol, benzeneethanol,4-ethoxy, acmc-1cit4, benzeneethanol, 4-ethoxy, 2-4-ethoxyphenyl ethan-1-ol
PubChem CID 89752
IUPAC Name 2-(4-ethoxyphenyl)ethanol
SMILES CCOC1=CC=C(CCO)C=C1

Specifications

Melting Point 40°C to 44°C
Boiling Point 138°C to 141°C (8 mmHg)
Quantity 2 g
Beilstein 3239377
Solubility Information Soluble in water.
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 2-(4-Ethoxyphenyl)ethanol
EINECSNumber 245-072-6
TSCA No

RUO – Research Use Only