missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diphenyloxazole 99.0+%, TCI America™

Catalog No. p-7126492
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09011G 1 g
D090125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09011G Supplier TCI America Supplier No. D09011G
Only null left

Chemical Identifiers

CAS 838-41-5
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.259
MDL Number MFCD00016890
InChI Key VUPXKQHLZATXTR-UHFFFAOYSA-N
PubChem CID 168750
IUPAC Name 2,4-diphenyl-1,3-oxazole
SMILES C1=CC=C(C=C1)C2=COC(=N2)C3=CC=CC=C3

Specifications

Melting Point 105°C
Color White
Boiling Point 340°C
Quantity 1 g
Formula Weight 221.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diphenyloxazole