accessibility menu, dialog, popup

UserName

CAS RN 838-41-5

ON

CAS RN 838-41-5

IUPAC Name: 2,4-diphenyl-1,3-oxazole
Synonyms: oxazole, 2,4-diphenyl-
Molecular Weight (g/mol): 221.26
Molecular Formula: C15H11NO
InChi Key: VUPXKQHLZATXTR-UHFFFAOYSA-N
SMILES: O1C=C(N=C1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results