missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diphenyl-6-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7128720
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52711G 1 g
D52715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52711G Supplier TCI America Supplier No. D52711G
Only null left

Chemical Identifiers

CAS 1219956-23-6
Molecular Formula C27H26BN3O2
Molecular Weight (g/mol) 435.334
InChI Key PVGOPEUJUVXCGN-UHFFFAOYSA-N
Synonym 4-(4,6-Diphenyl-1,3,5-triazine-2-yl)phenylboronic Acid Pinacol Ester, 2-[4-(4,6-Diphenyl-1,3,5-triazin-2-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 58340545
IUPAC Name 2,4-diphenyl-6-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=NC(=NC(=N3)C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 212°C
Color White
Quantity 1 g
Formula Weight 435.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diphenyl-6-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine