missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diphenyl-4-methyl-1-pentene 95.0+%, TCI America™

Catalog No. p-7127336
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D249725ML 25 mL
D2497100ML 100 mL
D2497500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D249725ML Supplier TCI America Supplier No. D249725ML
Only null left

Chemical Identifiers

CAS 6362-80-7
Molecular Formula C18H20
Molecular Weight (g/mol) 236.36
MDL Number MFCD00044027
InChI Key ZOKCNEIWFQCSCM-UHFFFAOYSA-N
Synonym 4-Methyl-2,4-diphenyl-1-pentene
PubChem CID 80715
IUPAC Name (4-methyl-4-phenylpent-1-en-2-yl)benzene
SMILES CC(C)(CC(=C)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 mL
UN Number 3082
Formula Weight 236.36
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Diphenyl-4-methyl-1-pentene
EINECSNumber (4)-0852&(4)-0854
TSCA Yes