missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride, 97%

Catalog No. p-7036526
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54833MD 250 mg
AAH5483303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54833MD Supplier Thermo Scientific Chemicals Supplier No. H54833MD

CAS: 56044-12-3 | C8H5ClN2O4S | 260.648 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56044-12-3
Molecular Formula C8H5ClN2O4S
Molecular Weight (g/mol) 260.648
MDL Number MFCD07186482
InChI Key LEFXQSDJQGOMPS-UHFFFAOYSA-N
Synonym 2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfonyl chloride, 2,4-dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride, 2,4-dioxo-1,3-dihydroquinazoline-6-sulfonyl chloride, 6-chlorosulfonyl-1,3-dihydroquinazoline-2,4-dione, pubchem5532, acmc-209lqm, 6-chlorosulfonylquinazoline-2,4-dione, 2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfony
PubChem CID 3162025
IUPAC Name 2,4-dioxo-1H-quinazoline-6-sulfonyl chloride
SMILES C1=CC2=C(C=C1S(=O)(=O)Cl)C(=O)NC(=O)N2

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 259.97
Percent Purity 97%
Chemical Name or Material 2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only