CAS RN 56044-12-3
IUPAC Name:
2,4-dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride
Synonyms:
2,4-dioxo-1,3-dihydroquinazoline-6-sulfonyl chloride
Molecular Weight (g/mol):
260.65
Molecular Formula:
C8H5ClN2O4S
InChi Key:
LEFXQSDJQGOMPS-UHFFFAOYSA-N
SMILES:
ClS(=O)(=O)C1=CC=C2NC(=O)NC(=O)C2=C1
1
–
1
of
1
results
1
Promotions Available
2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride, 97%
2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride, 97%
CAS: 56044-12-3 Molecular Formula: C8H5ClN2O4S Molecular Weight (g/mol): 260.648 MDL Number: MFCD07186482 InChI Key: LEFXQSDJQGOMPS-UHFFFAOYSA-N Synonym: 2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfonyl chloride,2,4-dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride,2,4-dioxo-1,3-dihydroquinazoline-6-sulfonyl chloride,6-chlorosulfonyl-1,3-dihydroquinazoline-2,4-dione,pubchem5532,acmc-209lqm,6-chlorosulfonylquinazoline-2,4-dione,2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfony PubChem CID: 3162025 IUPAC Name: 2,4-dioxo-1H-quinazoline-6-sulfonyl chloride SMILES: C1=CC2=C(C=C1S(=O)(=O)Cl)C(=O)NC(=O)N2