missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. D084625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D084625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D084625G Supplier TCI America Supplier No. D084625G

Chemical Identifiers

CAS 55907-61-4
Molecular Formula C6H7ClN4O4
Molecular Weight (g/mol) 234.60
MDL Number MFCD00050688
InChI Key PUYFAZQODQZOFK-UHFFFAOYSA-N
PubChem CID 5463312
IUPAC Name hydrogen (2,4-dinitrophenyl)hydrazine chloride
SMILES [H+].[Cl-].NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Quantity 25 g
UN Number 1325
Formula Weight 234.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitrophenylhydrazine Hydrochloride
EINECSNumber (3)-0478
TSCA No