missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenylacetic acid, 98%

Catalog No. p-7026217
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2210214 25 g
AAB2210222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2210214 Supplier Thermo Scientific Chemicals Supplier No. B2210214

CAS: 643-43-6 | C8H6N2O6 | 226.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 643-43-6
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.144
MDL Number MFCD00007227
InChI Key KCNISYPADDTFDO-UHFFFAOYSA-N
Synonym 2,4-dinitrophenylacetic acid, 2-2,4-dinitrophenyl acetic acid, 2,4-dinitrophenyl acetic acid, benzeneacetic acid, 2,4-dinitro, acmc-209nlg, 2,4-dimtrophenylacetic acid, ncistruc1_000234, ncistruc2_000110, 2,4-dinitrobenzeneacetic acid, benzeneacetic acid,4-dinitro
PubChem CID 221513
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 177°C to 182°C
Quantity 25 g
Beilstein 1990798
Formula Weight 226.15
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-398-2
RTECSNumber AH2230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only