missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenyl Thiocyanate 98.0+%, TCI America™

Catalog No. T01991G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01991G 1 g
T019925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01991G Supplier TCI America Supplier No. T01991G

Chemical Identifiers

CAS 1594-56-5
Molecular Formula C7H3N3O4S
Molecular Weight (g/mol) 225.178
MDL Number MFCD00047702
InChI Key XQDQRCRASHAZBA-UHFFFAOYSA-N
Synonym dntb, nirit, trirodazeen, gryzbol, grzybol, 2,4-dinitrothiocyanobenzene, nirit supra, tri-rodazene, 2,4-dinitrothiocyanatobenzene, rhodandinitrobenzol
PubChem CID 15325
ChEBI CHEBI:53051
IUPAC Name (2,4-dinitrophenyl) thiocyanate
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SC#N

Specifications

Melting Point 139°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 225.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitrophenyl Thiocyanate
RTECSNumber XK9450000
TSCA No