missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitroiodobenzene 98.0+%, TCI America™

Catalog No. D19215G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D19215G Supplier TCI America Supplier No. D19215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 709-49-9
Molecular Formula C6H3IN2O4
Molecular Weight (g/mol) 294.004
MDL Number MFCD00039738
InChI Key FXMKXMJLXRTQSW-UHFFFAOYSA-N
Synonym 2,4-dinitroiodobenzene, benzene, 1-iodo-2,4-dinitro, unii-not697x2xm, ccris 7907, 4-iodo-1,3-dintrobenzene, 6-iodo-1,3-dintrobenzene, 2,4-dinitro-1-iodobenzene, not697x2xm, 1-iodo-2,4-dinitro-benzene, 1-iod-2,4-dinitrobenzol
PubChem CID 69730
ChEBI CHEBI:59000
IUPAC Name 1-iodo-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])I

Specifications

Melting Point 87°C
Quantity 5 g
Formula Weight 294.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitroiodobenzene